missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Autoclave Bags, Cardboard holder for 511 bags

Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.

Brand:  Thermo Scientific™ S23B

Code : A0

Additional Details : Weight : 4.50000kg

Product Code. 12013072

  • 246.00 € / Pack of 10
Estimated Shipment: 14-05-2024
to see stock.
Add to basket

Description

Description

Thermo Scientific™ Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.

  • HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C
  • Convenient “tissue box” cartons dispense bags individually
  • Coated wire stands or cardboard holders available for use with autoclave bags

Ordering Alerts:

Not available in all geographic regions

Specifications

Specifications

Coated wire holder for 511 bags
Coated Wire
10
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers