missing translation for 'onlineSavingsMsg'
Learn More

Merck Steritop™ Threaded Bottle Top Filters

The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. Combine this Steritop™ filter unit with a receiver flask (sold separately) for processing and storing volumes from 150 to 1000 mL in a single, ready-to-use format.

Brand:  Merck S2GVT05RE

Additional Details : Weight : 2.14200kg

 View more versions of this product

Product Code. 16295122

  • 215.00 € / Pack of 12
Estimated Shipment: 02-04-2024
to see stock.
Add to basket

Description

Description

The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. Combine this Steritop™ filter unit with a receiver flask (sold separately) for processing and storing volumes from 150 to 1000 mL in a single, ready-to-use format.

  • Millipore Express™ PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs.
  • Durapore™ membrane for ultra-low protein binding for minimum absorption and protein loss.
  • Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient.
  • Sterilization of Aqueous Solutions (150 mL to 1 L)
    • Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component.
Specifications

Specifications

Clear with Blue Adapter
Polystyrene
0.22 μm
12
Radio sterilized
Hydrophilic
Durapore
PVDF
Sterile
45 mm
500 mL
12.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers