Learn More
Merck Steritop™ Threaded Bottle Top Filters
The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. Combine this Steritop™ filter unit with a receiver flask (sold separately) for processing and storing volumes from 150 to 1000 mL in a single, ready-to-use format.
137.00 € - 320.00 €
Specifications
Material | Polystyrene |
---|---|
Pore Size | 0.22 μm |
Sterility | Sterile |
Outlet Fitting | 45 mm |
Product Code | Brand | Material (Membrane) | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material (Membrane) | Volume (Metric) | Price | Quantity & Availability | ||||
15919180
|
Merck
S2GPT01RE |
PES | 150 mL |
137.00 €
Pack of 12 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
15929180
|
Merck
S2GPT02RE |
PES | 250 mL |
166.00 €
Pack of 12 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||||
15939180
|
Merck
S2GPT10RE |
PES | 1000 mL |
320.00 €
Pack of 12 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
16295122
|
Merck
S2GVT05RE |
PVDF | 500 mL |
215.00 €
Pack of 12 |
Estimated Shipment: 23-04-2024
Log in to see stock. |
||||
Description
The Stericup™ vacuum filtration system is the most efficient single-use, disposable Bottle Top Filter process for sterile media preparation. Combine this Steritop™ filter unit with a receiver flask (sold separately) for processing and storing volumes from 150 to 1000 mL in a single, ready-to-use format.
- Millipore Express™ PLUS membrane for fastest flow rates and low protein binding to minimize absorption and protein loss, which affect downstream performance and costs.
- Durapore™ membrane for ultra-low protein binding for minimum absorption and protein loss.
- Snug tight packaging, easy tear-open pouch, and convenient label designs make storing, opening and tracking your devices easy and efficient.
- Sterilization of Aqueous Solutions (150 mL to 1 L)
- Sterile Filtration of Culture Media and Buffers with High-Value Protein or Small Molecule Component.
Specifications
Polystyrene | |
0.22 μm | |
Sterile | |
45 mm |