missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Storage File Systems

For long-term storage of histology microscope slide and tissue cassette specimens.

186.00 €

Specifications

Color White
For Use With (Application) Histology Speciemen Storage
Height (Metric) 100 mm
Material Cardboard
Width (Metric) 299 mm
View More Specs

Products 1
Product Code Brand Device Holds Price Quantity & Availability  
Product Code Brand Device Holds Price Quantity & Availability  
17207820
View Documents Promotion Details
Fisherbrand™
500 Tissue Block
186.00 €
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 10-07-2024
to see stock.
Add to basket
 
Description

Description

Fisherbrand™ slide and tissue cassette storage boxes provide an economical yet sturdy and secure method for long-term storage of your microscope slide and tissue cassette block specimens.

Our easy to assemble boxes come 10 per case and include 40 drawers/drawer inserts for the slide storage option (4 drawers per file) and 80 drawers/drawer inserts for the tissue block storage option (8 drawers per file).

The coated surface helps to ensure moisture resistance for long-term protection. Each microscope slide storage box holds up to 2000 microscope slides while each tissue block storage box holds up to 500 tissue blocks/cassettes. The unique locking tabs help keep stacked boxes safely secured and stable.

Specifications

Specifications

White
100 mm
299 mm
Histology Speciemen Storage
Cardboard
Videos
SDS
Documents

Documents

Product Certifications