missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Straight Wall Inconel Crucible

Brand:  Alfa Aesar™ 036012.KT

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15476145

  • 87.20 € / Each


Estimated Shipment date
from Supplier 20-06-2019
Add to basket

Specifications

Specifications

55mL
Outside: 46mm
Inconel
Straight Wall
Inconel Crucible
Videos
SDS