Learn More
Thermo Scientific Chemicals Straight Wall Nickel Crucible
22.70 € - 245.00 €
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
15456135
|
Thermo Scientific™
035889.KT |
5 mL | Outside: 21mm |
45.30 €
Each |
In Stock
Log in to see stock. |
||||
15416145
|
Thermo Scientific Chemicals
035952.KT |
20 mL | Outside: 33mm |
34.30 €
Each |
In Stock
Log in to see stock. |
||||
15436145
|
Thermo Scientific™
035973.KT |
25 mL | Outside: 45mm |
49.70 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
15466145
|
Thermo Scientific Chemicals
035991.KT |
35 mL | Outside: 46mm |
48.10 €
Each |
Estimated Shipment: 27-05-2024
Log in to see stock. |
||||
15486145
|
Thermo Scientific™
036013.KT |
55 mL | Outside: 46mm |
58.80 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
15406155
|
Thermo Scientific Chemicals
036053.KT |
100 mL | Outside: 59mm |
100.00 €
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
||||
15416155
|
Thermo Scientific Chemicals
036071.KT |
55 mL | Outside: 47mm |
61.20 €
Each |
In Stock
Log in to see stock. |
||||
15436155
|
Thermo Scientific Chemicals
036088.KT |
500 mL | Outside: 101mm |
245.00 €
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
15446155
|
Thermo Scientific Chemicals
036098.KT |
250 mL | Outside: 82mm |
143.00 €
Each |
In Stock
Log in to see stock. |
||||
15476135
|
Thermo Scientific™
035910.KT |
10 mL | Outside: 27mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15496135
|
Thermo Scientific™
035931.KT |
15 mL | Outside: 33mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15466155
|
Thermo Scientific™
036112.KT |
75 mL | Outside: 51mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||