Learn More
IBA Lifesciences Strep-Tactin™XT Superflow™ High Capacity Resin
Strep-Tactin™XT Superflow™ high capacity resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
Brand: IBA Lifesciences 2-4030-025
Code : NEW
Additional Details : Weight : 0.04500kg
Description
- Strep-Tactin™XT Superflow™ high capacity is a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
- It allows the addition of denaturing agents such as urea during the purification procedure. In comparison to Strep-Tactin™ XT Superflow™, Strep-Tactin™XT Superflow™ high capacity has a higher density of immobilized Strep-Tactin™XT, which leads to a 3- to 5-fold higher yield of purified protein.
- As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein.
- The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
- To pack a column with 1 ml resin bed volume, 2 ml 50% suspension are necessary.
Specifications
50 mL | |
High Capacity Resin |
For Research Use Only.