missing translation for 'onlineSavingsMsg'
Learn More

Strontium bis(trifluoromethylsulfonyl)imide, Thermo Scientific™

119.00 € - 486.00 €

Chemical Identifiers

CAS 862121-57-1
Molecular Formula C4F12N2O8S4Sr
Molecular Weight (g/mol) 647.891
MDL Number MFCD23380176
InChI Key DIMYICIXZTVMOY-UHFFFAOYSA-N
Synonym Bis(trifluoromethane)sulfonimide strontium salt; Bis(trifluoromethanesulfonyl)imide strontium salt
PubChem CID 134159298
IUPAC Name strontium;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Sr+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15472837
View Documents
Thermo Scientific Alfa Aesar
047099.03
1g
119.00 €
1g
View alternative products
Save Up to 
»
In Stock
Add to basket
 
 Restricted Item
15482837
View Documents
Thermo Scientific Alfa Aesar
047099.06
5g
486.00 €
5g
View alternative products
Save Up to 
»
Estimated Shipment: 26-08-2026
Add to basket
 
 Restricted Item
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

862121-57-1
647.891
DIMYICIXZTVMOY-UHFFFAOYSA-N
134159298
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Sr+2]
C4F12N2O8S4Sr
MFCD23380176
Bis(trifluoromethane)sulfonimide strontium salt; Bis(trifluoromethanesulfonyl)imide strontium salt
strontium;bis(trifluoromethylsulfonyl)azanide

Specifications

862121-57-1
[(CF3SO2)2N]2Sr
UN3261
DIMYICIXZTVMOY-UHFFFAOYSA-N
strontium;bis(trifluoromethylsulfonyl)azanide
134159298
Hygroscopic
C4F12N2O8S4Sr
MFCD23380176
Bis(trifluoromethane)sulfonimide strontium salt; Bis(trifluoromethanesulfonyl)imide strontium salt
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Sr+2]
647.891
647.91
Strontium bis(trifluoromethylsulfonyl)imide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.