Learn More
Techne™ Electronic Ice Bucket Ceramic Beads
Keep your samples cold, clean, and dry
Brand: Techne™ 24120-99
Code : 57
Additional Details : Weight : 0.80000kg
Description
These Techne™ N°ICE ceramic-coated beads maintain the temperature and support the sample vessels for as long as necessary. The beads are chemically resistant and if required can be autoclaved at 134°C. As a time saving option, the bucket of beads can be incubated in the fridge overnight and simply inserted into the unit as and when required. Compatible with the 24120-00 Electronic Ice Bucket, sold separately.
Specifications
Replacement Ceramic Beads |