missing translation for 'onlineSavingsMsg'
Learn More

Styrene, 99.5%, for analysis, stabilized, Thermo Scientific Chemicals

Catalog Number p-194017
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
100-42-5, 98-29-3
C8H8
104.15
MFCD00008612,MFCD00084450
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
7501
CHEBI:27452
styrene
C=CC1=CC=CC=C1
100-42-5, 98-29-3
C8H8
104.15
MFCD00008612,MFCD00084450
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
7501
CHEBI:27452
styrene
C=CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -31.0°C
Density 0.9060g/mL
Boiling Point 145.0°C to 146.0°C
Flash Point 31°C
Infrared Spectrum Authentic
Assay Percent Range 99.4% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH=CH2
Refractive Index 1.5460 to 1.5480
Quantity 250 mL
Beilstein 05, 474
Merck Index 15, 8990
Specific Gravity 0.906
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: soluble in alcohol,ether,acetone,cs2,ccl4,dissolves organic substances and polymers
Stabilizer 10 to 20ppm p-tert-butylcatechol
Viscosity 0.695 mPa.s (25°C)
Formula Weight 104.15
Percent Purity 99.5%
Grade Analysis
Physical Form Liquid
Chemical Name or Material Styrene, p.a.
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Styrene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H372-Causes damage to organs.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'