Learn More
Sucrose, Extra Pure, SLR, Fisher Chemical™
Brand: Fisher Chemical S/8560/53
Code : 21
Additional Details : CAS Number : 57-50-1 Weight : 0.50000kg
Packaging | Plastic powder jar |
---|---|
Quantity | 500g |
Chemical Identifiers
57-50-1 | |
342.297 | |
CZMRCDWAGMRECN-UGDNZRGBSA-N | |
5988 | |
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
C12H22O11 | |
6626 | |
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand | |
CHEBI:17992 | |
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O |
Specifications
Plastic powder jar | |
190°C | |
6.5 to 7.5 | |
C12H22O11 | |
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand | |
C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O | |
342.297 | |
CHEBI:17992 | |
Solid |
White | |
500g | |
57-50-1 | |
6626 | |
CZMRCDWAGMRECN-UGDNZRGBSA-N | |
(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
5988 | |
342.29g/mol |