missing translation for 'onlineSavingsMsg'
Learn More

Sucrose, Ultrapure Bioreagent, J.T. Baker™

Catalog Number p-5391275
102.60 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
57-50-1
C12H22O11
342.30
MFCD00006626
CZMRCDWAGMRECN-PWPRYFECNA-N
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
5988
CHEBI:17992
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
This item is not returnable. View return policy
57-50-1
C12H22O11
342.30
MFCD00006626
CZMRCDWAGMRECN-PWPRYFECNA-N
sucrose, saccharose, cane sugar, sugar, table sugar, white sugar, d-sucrose, saccharum, rohrzucker, amerfand
5988
CHEBI:17992
OC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Density Gradient Centrifugation and Molecular Biology Applications
Packaging Poly Bottle
Quantity 1 kg
Specific Gravity 1L = 1.59kg
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Formula Weight 342.3
Grade Ultrapure Bioreagent
Chemical Name or Material Sucrose
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'