missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sucrose, Ultrapure Bioreagent, J.T. Baker™
102.60 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Description
- For Density Gradient Centrifugation and Molecular Biology Applications
Specifications
Packaging | Poly Bottle |
Quantity | 1 kg |
Specific Gravity | 1L = 1.59kg |
IUPAC Name | (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
Formula Weight | 342.3 |
Grade | Ultrapure Bioreagent |
Chemical Name or Material | Sucrose |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'