missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamic acid, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Melting Point | 200.0°C to 208.0°C |
Packaging | Plastic Bottle |
Assay Percent Range | 99% |
Quantity | 250 g |
Linear Formula | H2NSO3H |
Physical Form | Crystals or Crystalline Powder |
Fieser | 11117 |
Merck Index | 159053 |
Solubility Information | Solubility in water: 146.8g/L (20°C). Other solubilities: sparingly soluble in alcohol and methanol,slightly soluble in acetone,insoluble in ether |
Show More |
Safety and Handling
Product Identifier
- Sulfamic acid
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'