missing translation for 'onlineSavingsMsg'
Learn More

Sulfisoxazole, 99%, Thermo Scientific Chemicals

Catalog Number p-4813671
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
127-69-5
C11H13N3O3S
267.3
NHUHCSRWZMLRLA-UHFFFAOYSA-N
sulfisoxazole, sulfafurazole, sulphafurazole, sulfisoxazol, sulfafurazol, sulfaisoxazole, sulfofurazole, sulfisoxasole, sulphaisoxazole, sulfadimethylisoxazole
5344
CHEBI:102484
4-amino-N-(3,4-dimethyl-1,2-oxazol-5-yl)benzenesulfonamide
CC1=C(ON=C1C)NS(=O)(=O)C2=CC=C(C=C2)N
This item is not returnable. View return policy
127-69-5
C11H13N3O3S
267.3
NHUHCSRWZMLRLA-UHFFFAOYSA-N
sulfisoxazole, sulfafurazole, sulphafurazole, sulfisoxazol, sulfafurazol, sulfaisoxazole, sulfofurazole, sulfisoxasole, sulphaisoxazole, sulfadimethylisoxazole
5344
CHEBI:102484
4-amino-N-(3,4-dimethyl-1,2-oxazol-5-yl)benzenesulfonamide
CC1=C(ON=C1C)NS(=O)(=O)C2=CC=C(C=C2)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 194°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 15,9082
Solubility Information Solubility in water: 0.13mg/mL (25°C). Other solubilities: soluble in alcohol
Formula Weight 267.3
Percent Purity 99%
Chemical Name or Material Sulfisoxazole
compliance-icons
Product Identifier
  • Sulfisoxazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'