missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Oil Standards for Diesel Fuel Analysis, SPEX CertiPrep™

Catalog Number p-4279733
Click to view available options
Concentration:
10 μg/g
100 μg/g
1000 μg/g
200 μg/g
25 μg/g
300 μg/g
5 μg/g
50 μg/g
75 μg/g
750 μg/g
This item is not returnable. View return policy
7704-34-9, 8042-47-5
S
32.06
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
5362487
CHEBI:26833
sulfur
[S]
This item is not returnable. View return policy
7704-34-9, 8042-47-5
S
32.06
MFCD00085316
NINIDFKCEFEMDL-UHFFFAOYSA-N
5362487
CHEBI:26833
sulfur
[S]

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
S5μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Physical Form Solid
CAS Min % 0.0, 99.99
CAS Max % 0.0, 99.99
Concentration 5 μg/g
Packaging Triple-leached LDPE bottle
Solvent or Matrix Base Oil 20
Quantity 100 mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Chemical Name or Material Sulfur Oil Standard in Base Oil
compliance-icons
Product Identifier
  • Sulfur Oil Standard in Base Oil 20
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Mineral Oil

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'