missing translation for 'onlineSavingsMsg'
Learn More

Sulfur Oil Standards for Diesel Fuel Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep DSS8-25Y

Code : A0

Additional Details : Weight : 0.14000kg

Concentration 25 μg/g

 View more versions of this product

Product Code. 11945094

  • 136.49 € / 100g

Estimated Shipment
27-05-2021

Please to purchase this item. Need a web account? Register with us today!


Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
S25μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7704-34-9
32.06
5362487
sulfur
S
NINIDFKCEFEMDL-UHFFFAOYSA-N
CHEBI:26833
[S]

Specifications

Sulfur Oil Standard in Base Oil
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
99.99
S
NINIDFKCEFEMDL-UHFFFAOYSA-N
sulfur
5362487
Solid
Base Oil 20
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
7704-34-9,8042-47-5
99.99
25 μg/g
[S]
32.06
CHEBI:26833
Triple-leached LDPE bottle
100mL
Videos
SDS
Documents

Product intended for laboratory use only