Learn More
Sulfur Oil Standards for Diesel Fuel Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep DSS8-75Y
Code : A0
Additional Details : Weight : 0.14000kg
Concentration | 75 μg/g |
---|
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
S | 75μg/g |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7704-34-9 | |
32.06 | |
5362487 | |
sulfur |
S | |
NINIDFKCEFEMDL-UHFFFAOYSA-N | |
CHEBI:26833 | |
[S] |
Specifications
Sulfur Oil Standard in Base Oil | |
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation | |
99.99 | |
S | |
NINIDFKCEFEMDL-UHFFFAOYSA-N | |
sulfur | |
5362487 | |
Solid | |
Base Oil 20 |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
7704-34-9,8042-47-5 | |
99.99 | |
75 μg/g | |
[S] | |
32.06 | |
CHEBI:26833 | |
Triple-leached LDPE bottle | |
100mL |
Product intended for laboratory use only