missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Min 95% d=1.83, Certified AR, for Analysis, Fisher Chemical™
24.42 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
42.56 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "21741" to get your promotional price.
42.56 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic Coated Glass bottle
Plastic container
Description
H2O4S, CAS Number-7664-93-9, electrolyte acid, acidum sulfuricum, battery acid, dihydrogen sulfate, sulphuric acid, oil of vitriol, acide sulfurique, mattling acid, vitriol brown oil, dipping acid, 2.5L, 290 deg.C, CHEBI:26836, Colorless, 98.07g/mol, QAOWNCQODCNURD-UHFFFAOYSA-N, sulfuric acid
Specifications
pH | 1 |
Color | Colorless |
Melting Point | 10°C |
Boiling Point | 290°C |
Packaging | HDPE plastic bottle |
Vapor Pressure | 1mmHg 146 |
Quantity | 1 L |
Viscosity | 21mPa at 25°C |
Physical Form | Liquid |
Formula Weight | 98.07g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'