missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Solution, 49 to 51%, For HPLC, Honeywell Fluka™

Catalog Number p-7120850
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7664-93-9
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7664-93-9
H2O4S
98.07
MFCD00064589
QAOWNCQODCNURD-UHFFFAOYSA-N
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
1118
CHEBI:26836
sulfuric acid
OS(O)(=O)=O

for HPLC, 49-51%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
pH Acidic
Color Colorless
Boiling Point 110°C
Density 1.403g/mL at 20°C
Odor Odorless
Packaging Glass bottle
Quantity 100 mL
Linear Formula H2SO4
UN Number UN2796
Beilstein 2037554
Formula Weight 98.08g/mol
Percent Purity 49 to 51%
Chemical Name or Material Sulfuric acid solution
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'