Learn More
Thermo Scientific™ Super Clean™ Gas Cartridge Filters
Improve gas quality and optimize GC analysis with easy-to-use Thermo Scientific™ Super Clean™ Gas Cartridge Filters.
Brand:  Thermo Scientific™ 60180-832
Code : 16
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10427144
  • 1085.00 € / Each
Estimated Shipment
21-03-2019
Add to basket
Specifications
Specifications
Inert Carrier, He, H2, N2, AR, Air
Inert Carrier, He, H2, N2, AR, Air
7.2 g H2O
Twin Moisture Filters for High Flow (Air Generators)
Moisture
Baseplate not included
Videos
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.