Learn More
Metro™ Super Erecta™ Wire Shelf, Stainless Steel Finish
Transform your laboratory or workplace storage as you see fit. Metro Super Erecta Wire Shelves in stainless steel finish allow adjustable configurations when used with Metro Super Erecta shelving systems
144.00 € - 715.00 €
Specifications
For Use With (Equipment) | Metro™ Super Erecta™ shelving system |
---|---|
Includes | Plastic sleeves and wedges |
Material | Stainless Steel |
Product Code | Brand | Dimensions (L x W) | Depth (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Depth (Metric) | Price | Quantity & Availability | ||||
15946611
|
Metro™
1872NS |
45.7 x 183 in. (18 x 72 cm) | 457 mm |
413.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15966681
|
Metro™
3048NS |
76 x 121.9 in. (30 x 48 cm) | 760 mm |
455.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15946681
|
Metro™
3036NS |
76 x 91.4 in. (30 x 36 cm) | 760 mm |
390.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15916671
|
Metro™
2448NS |
61 x 121.9 in. (24 x 48 cm) | 610 mm |
344.00 €
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
||||
15926591
|
Metro™
1824NS |
45.7 x 61 in. (18 x 24 cm) | 457 mm |
229.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15976671
|
Metro™
2460NS |
61 x 152.4 in. (24 x 60 cm) | 610 mm |
419.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15966601
|
Metro™
1848NS |
45.7 x 121.9 in. (18 x 48 cm) | 457 mm |
274.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15936661
|
Metro™
2436NS |
61 x 91.4 in. (24 x 36 cm) | 610 mm |
218.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15906601
|
Metro™
1836NS |
45.7 x 91.4 in. (18 x 36 cm) | 457 mm |
176.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15916571
|
Metro™
1430NS |
35.5 x 76 in. (14 x 30 cm) | 355 mm |
144.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15906581
|
Metro™
1448NS |
35.5 x 121.9 in. (14 x 48 cm) | 355 mm |
176.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15956581
|
Metro™
1460NS |
35.5 x 152.4 in. (14 x 60 cm) | 355 mm |
222.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15986581
|
Metro™
1472NS |
35.5 x 182.9 in. (14 x 72 cm) | 355 mm |
328.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15996611
|
Metro™
2124NS |
53 x 63 in. (28 x 28 cm) | 530 mm |
266.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15986631
|
Metro™
2148NS |
53 x 21.9 in. (21 x 48 cm) | 530 mm |
308.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15946621
|
Metro™
2130NS |
53 x 76 in. (30 x 30 cm) | 530 mm |
268.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15936631
|
Metro™
2142NS |
53 x 106.6 in. (21 x 42 cm) | 530 mm |
308.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15916641
|
Metro™
2154NS |
53 x 37 in. (21 x 54 cm) | 530 mm |
312.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15996641
|
Metro™
2172NS |
53 x 182.9 in. (21 x 72 cm) | 530 mm |
370.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15956641
|
Metro™
2160NS |
53 x 152.4 in. (21 x 60 cm) | 530 mm |
312.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15986661
|
Metro™
2442NS |
61 x 106.6 in. (24 x 42 cm) | 610 mm |
344.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15936651
|
Metro™
2424NS |
61 x 61 in. (24 x 24 cm) | 610 mm |
215.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15946671
|
Metro™
2454NS |
61 x 137 in. (24 x 54 cm) | 610 mm |
419.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15926691
|
Metro™
3072NS |
76 x 182.9 in. (30 x 72 cm) | 760 mm |
615.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15916701
|
Metro™
3636NS |
91 x 91 in. (36 x 36 cm) | 910 mm |
438.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15986701
|
Metro™
3672NS |
91 x 182.9 in. (36 x 72 cm) | 910 mm |
715.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15956701
|
Metro™
3660NS |
91 x 152.4 in. (36 x 60 cm) | 910 mm |
600.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
Description
- Open-wire design minimizes dust accumulation, allows free circulatin of air and greater visibility of stored items
- Type 304 polished stainless steel
- Used for the most aggressive applications and environments
- Stands up to high temperature automated cart wash and autoclave applications
Specifications
Metro™ Super Erecta™ shelving system | |
Plastic sleeves and wedges | |
Stainless Steel |