Learn More
Thermo Scientific™ SuperBlock™ Blocking Buffer, blotting
Thermo Scientific SuperBlock Blocking Buffer, blotting is an optimized PBS solution containing a protein that provides incredibly fast and effective blocking for western blot analysis.
Brand: Thermo Scientific™ 37537
Code : A0
Additional Details : Weight : 1.00000kg
Description
Thermo Scientific SuperBlock Blocking Buffer, blotting is an optimized PBS solution containing a protein that provides incredibly fast and effective blocking for western blot analysis. Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems.
Features of SuperBlock Blocking Buffer, blotting include:• Fast—block membranes in 5 to 10 minutes
• Flexible—guaranteed to be biotin-free for use with streptavidin system
• Low background—the non-serum protein solution yields a high signal-to-noise ratio
• Stable—store buffer at 4°C for one year; store blocked plates dry for up to 12 months
Blocking micro-plates, membranes, or tissues with SuperBlock buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.
Specifications
SuperBlock (TBS) Blocking Buffer, blotting | |
1 L | |
Western Blot | |
1X | |
Single purified protein |
Blocking Buffer | |
Liquid | |
SuperBlock™ | |
TBS | |
Upon receipt store at 4°C. Product is shipped at ambient temperature. |