Learn More
Cytiva (formerly Pall Lab) Supor™ 100 Membrane Disc Filters
Membranes are used to sterilize liquid reagents, remove particulate contamination, and clarify solutions prior to further processing.
Brand:  Cytiva (formerly Pall Lab) 60310
Additional Details : Weight : 0.11400kg
 View more versions of this product
Product Code. 17104511
  • 227.00 € / Pack of 100
Estimated Shipment: 26-06-2024
to see stock.
Add to basket
Description
Description
  • These products are optimized for biological, pharmaceutical and sterilizing filtration requirements
  • For research use only
Specifications
Specifications
PES
Non-sterile
Plain
0.1 μm
132 μm
47 mm