missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Supor™ 100 Membrane Disc Filters
Membranes are used to sterilize liquid reagents, remove particulate contamination, and clarify solutions prior to further processing.
Brand: Cytiva (formerly Pall Lab) 66551
Additional Details : Weight : 0.49400kg
Description
- These products are optimized for biological, pharmaceutical and sterilizing filtration requirements
- For research use only
Specifications
PES | |
Non-sterile | |
Plain |
0.1 μm | |
132 μm | |
142 mm |