Learn More
Cytiva (formerly Pall Lab) Supor™ 100 Membrane Disc Filters
Membranes are used to sterilize liquid reagents, remove particulate contamination, and clarify solutions prior to further processing.
Brand:  Cytiva (formerly Pall Lab) 66552
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 17174571
  • 670.00 € / Pack of 25
Estimated Shipment: 08-11-2024
to see stock.
Add to basket
Description
Description
  • These products are optimized for biological, pharmaceutical and sterilizing filtration requirements
  • For research use only
Specifications
Specifications
PES
Non-sterile
Plain
0.1 μm
132 μm
293 mm