Learn More
Cytiva (formerly Pall Lab) Supor™ Membrane Disc Filters
Membranes are used to sterilize liquid reagents, remove particulate contamination, and clarify solutions prior to further processing.
Brand:  Cytiva (formerly Pall Lab) 61854
Additional Details : Weight : 0.24700kg
Product Code. 17134531
  • 212.00 € / Pack of 100
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • Optimized for biological, pharmaceutical and sterilizing filtration requirements
Specifications
Specifications
PES
Sterile
140 μm
47 mm
0.45 μm
100/Pk.
Gridded