Learn More
OHAUS™ Support Stand
OHAUS™ Support Stand provides secure support for holding glassware at elevated positions.
Brand:  OHAUS™ 30392338
Additional Details : Weight : 6.80000kg
 View more versions of this product
Product Code. 15722389
  • 433.00 € / Each
Estimated Shipment: 29-08-2024
to see stock.
Add to basket
Description
Description
  • Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
  • Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
  • Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
Specifications
Specifications
Support Stand with Rod
Clamp, Support, Stand Rod, CLR-STRODC102
305 mm
305.00 mm
16.50 in.
Stainless Steel
16 mm
12.00 in.
1016 mm
419.00 mm