Learn More
Fisherbrand™ SureOne™ Specialty Pipettor Tips, Filtered
Choose from extended length or large orifice, sterile or non-sterile. Fisherbrand™ Specialty Pipettor Tips, Filtered are certified to be free of RNase, DNase and DNA.
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 1.35000kg
Description
- Extended length tips and large orifice tips formats available
- Non-sterile items are certified to be free of RNase/DNase and DNA
- Sterile items are e-beam sterile and certified to be free of RNase/ DNase, pyrogen, bioburden, PCR* inhibitors and endtoxins
- Compatible with most popular brands of pipettor (optimal fit with Fisherbrand Elite™ pipettor)
Specifications
20 to 200 μL | |
Sterile | |
50 mm | |
Low Retention | |
Genomic/Wide Orifice | |
SureOne |
Filtered | |
Clear | |
DNase, RNase, ATP, Bioburden, PCR Inhibitor, Endotoxin, and Pyrogen Free | |
Rack | |
No |