Learn More
SVL™ Bearing Stirrer for SVL™ Stirrer Shaft
Designed to accept SVL™ stirrer shafts with ground glass surfaces.
Brand:  SVL 588-08
Code : NEW
Additional Details : Weight : 0.07000kg
 View more versions of this product
Product Code. 15547730
  • 890.00 € / Pack of 5
Call for latest local availability
01 885 5854
Add to basket
Description
Description
  • Borosilicate glass complete with fittings
  • Glands manufactured from PTFE based polymer which is self lubricating and corrosion-resistant
  • Two types available: With SVL™ screwthread to allow connection to SVL™ flasks, With Quickfit™ ground glass cone to allow connection to Quickfit™ flasks
Specifications
Specifications
SVL™ stirrer shaft
Quickfit™ ground Glass cone to allow connection to Quickfit™ flask
PTFE Based Polymer
24/29