Learn More
SVL™ Bearing Stirrer for SVL™ Stirrer Shaft
Designed to accept SVL™ stirrer shafts with ground glass surfaces.
283.00 € - 1465.00 €
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
11937298
|
SVL
588-09 |
29/32 |
283.00 €
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
15537730
|
SVL
588-07 |
19/26 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15527730
|
SVL
588-03 |
30 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15557730
|
SVL
588-12 |
45/40 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Borosilicate glass complete with fittings
- Glands manufactured from PTFE based polymer which is self lubricating and corrosion-resistant
- Two types available: With SVL™ screwthread to allow connection to SVL™ flasks, With Quickfit™ ground glass cone to allow connection to Quickfit™ flasks