Learn More
Quickfit™ Claisen Still Head Adapter
PTFE joint sleeves eliminate the possibility of joint seizure
Brand: Quickfit SH6/23
Code : 01
Additional Details : Weight : 0.09000kg
Description
- Similar to the SH4: still heads but with extra socket to accept dropping funnel or stirrer
Specifications
Claisen Still Head | |
Borosilicate Glass | |
19/26 (Condenser), 24/29 (Flask) | |
Socket: 14/23 |