Learn More
Quickfit™ Claisen Still Head Adapter
PTFE joint sleeves eliminate the possibility of joint seizure
Brand: Quickfit SH6/1
Code : NEW
Additional Details : Weight : 0.06000kg
Description
Similar to the SH4 still heads but with extra socket to accept dropping funnel or stirrer etc.Specifications
14/23 | |
Used for thermometer, dropping funnel, stirrer | |
Borosilicate Glass | |
Claisen still head Adapter |