missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Swainsonine

Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Inhibitor of α-mannosidase II which inhibits glycoprotein processing. Displays anticancer and immune modulatory properties.
Specifications
Specifications
| CAS | 72741-87-8 |
| Quantity | 1 mg |
| Molecular Formula | C8H15NO3 |
| Formula Weight | 173.21 |
| Purity | >98% |
| Molecular Weight (g/mol) | 173.21 |
| Chemical Name or Material | Swainsonine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction