missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Syk Inhibitor
Click to view available options
Quantity:
5 mg
Description
A cell-permeable oxindole compound that acts as a potent, reversible, and ATP-competitive Syk inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 5 mg |
Description | Dark orange |
Content And Storage | 2°C to 8°C |
For Use With (Application) | A cell-permeable oxindole compound that acts as a potent, reversible, and ATP-competitive Syk inhibitor |
Formula Weight | 353.40 |
Sensitivity | Light Sensitive |
Solubility | Soluble in DMSO |
Form | Solid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'