missing translation for 'onlineSavingsMsg'
Learn More

sym-Diphenylcarbazide, 98%, ACS reagent, Thermo Scientific Chemicals

Catalog Number 10427000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
140-22-7
C13H14N4O
242.28
KSPIHGBHKVISFI-UHFFFAOYSA-N
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
8789
CHEBI:4641
1,3-dianilinourea
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2
140-22-7
C13H14N4O
242.28
KSPIHGBHKVISFI-UHFFFAOYSA-N
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl
8789
CHEBI:4641
1,3-dianilinourea
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
Melting Point 173.0°C to 176.0°C
Color Beige to Pink
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5 % min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 15,292
Sensitivity (to chromate) passes test
Merck Index 15,3352
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in glacial acetic acid,soluble in alcohol and acetone
Formula Weight 242.28
Percent Purity 98%
Grade ACS Reagent
Physical Form Powder or Flakes
Chemical Name or Material sym-Diphenylcarbazide
Show More Show Less
compliance-icons
Product Identifier
  • sym-Diphenylcarbazide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'