missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Syringe Filters
27.20 € - 28.10 €
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10025671
|
Macherey-Nagel™
729100 |
2 mL |
27.20 €
Pack of 100 |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
10660732
|
Macherey-Nagel™
729101 |
5 mL |
28.10 €
Pack of 100 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Specifications
Single-use | |
100 Pack | |
Syringe Filter |