Learn More
Trajan Scientific and Medical™ PerkinElmer Series 200 LC Autosampler Syringe
Accurate and reproducible injections.
Brand: Trajan Scientific and Medical™ 007995
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for PerkinElmer instruments
Specifications
PTFE (Plunger) | |
High-performance Liquid Chromatography | |
500 μL | |
Gas Tight |