Learn More
SYT8 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20671-1-AP-150UL
 View more versions of this product
Product Code. 18078105
  • 410.00 € / 150µL
Description
The SYT8 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human SYT8. This antibody recognizes human, mouse antigen. The SYT8 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
SYT8
300 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, Immunofluorescence, ELISA
Synaptotagmin 8, synaptotagmin VIII, SYT8, SytVIII
150 μL
90019
Store at -20°C. Stable for one year after shipment.