Learn More
IBA Lifesciences TACS Column Adapter
Adapter for Strep-Tactin™ TACS Agarose Columns that fits onto standard tubes
Brand:  IBA Lifesciences 6-6331-001
Additional Details : Weight : 0.05000kg
 View more versions of this product
Product Code. 15819794
  • 10.00 € / Each
Estimated Shipment: 25-06-2024
to see stock.
Add to basket
Description
Description
  • The adapters allow fitting of Strep-Tactin™ TACS Agarose columns onto standard tubes.
  • Column adapters are available in two sizes: For 3 ml columns (containing 0.3 ml Strep-Tactin™ TACS Agarose) to fit them onto 15 ml tubes and for 10 ml columns (containing 1 ml Strep-Tactin™ TACS Agarose) to fit them onto 50 ml tubes. 
Specifications
Specifications
One adapter
1 mL