missing translation for 'onlineSavingsMsg'
Learn More

Tantalum powder, 99.98% (metal basis), Alfa Aesar™, Puratronic™

Nb 50ppm

Brand:  Alfa Aesar 010344.88

Code : NEW

Additional Details : CAS Number : 7440-25-7 Weight : 0.00250kg Transport : UN number : 3089 Chem class : 4.1 Pack group : III

Quantity 2.5g

 View more versions of this product

Product Code. 15475935

  • 26.30 € / 2.50g

EU Stock 7

Please to purchase this item. Need a web account? Register with us today!


Specifications

Chemical Identifiers

7440-25-7
180.948
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]

Specifications

Tantalum
7440-25-7
99.98% (Metals basis)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
180.948
CHEBI:33348
Black to Gray
2.5g
Gray to black powder
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
Odorless
23956
Powder
Videos
SDS