missing translation for 'onlineSavingsMsg'
Learn More

Tantalum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.98% (metals basis), Thermo Scientific Chemicals

Catalog Number 15464725
Click to view available options
Quantity:
1 Pc.
5 Pc.
This item is not returnable. View return policy
7440-25-7
Ta
180.95
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
23956
CHEBI:33348
tantalum
[Ta]
This item is not returnable. View return policy
7440-25-7
Ta
180.95
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
23956
CHEBI:33348
tantalum
[Ta]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 Pc.
Percent Purity 99.98%
Odor Odorless
Weight ≈6.8g/slug
Form Slug
Assay (metals basis)
Chemical Name or Material Tantalum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'