missing translation for 'onlineSavingsMsg'
Learn More

Tantalum tubing, 3.18mm (0.125in) OD, 2.26mm (0.089in) ID, 99.9% (metals basis), Thermo Scientific™

Catalog Number 15474915
Click to view available options
Quantity:
10 cm
50 cm
This item is not returnable. View return policy
7440-25-7
Ta
180.95
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
23956
CHEBI:33348
tantalum
[Ta]
This item is not returnable. View return policy
7440-25-7
Ta
180.95
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
23956
CHEBI:33348
tantalum
[Ta]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 10 cm
Percent Purity 99.9%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Tantalum tubing, OD: 3.18mm (0.125 in.), ID: 2.26mm (0.089 in.)
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'