missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals TAPI-2
Click to view available options
Quantity:
1 mg
Description
A hydroxamate-based inhibitor of MMPs and TACE
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 1 mg |
Description | White |
Content And Storage | -30°C to -10°C |
For Use With (Application) | A hydroxamate-based inhibitor of MMPs and TACE |
CAS | 187034-31-7 |
Molecular Formula | C19H37N5O5 |
Formula Weight | 415.53 |
Synonym | TNF-β Protease Inhibitor-2; |
Sensitivity | Light Sensitive and Hygroscopic |
Form | Solid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'