missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Protein Expression Assay Kit (hICAM1)
TaqMan™ Protein Expression Assay Kit (hICAM1)
Brand: Applied Biosystems™ 4405471
Additional Details : Weight : 0.01000kg
Description
Applied Biosystems™ TaqMan™ Protein Expression Assays enable fast, easy identification, and relative protein quantification of pluripotency protein markers from limited quantities of cultured hESCs. The TaqMan™ Protein Expression Assay Kit (Human ICAM1) contains the assay probes and a dilution buffer for the labelling of human inter-cellular adhesion molecule 1 protein (hICAM1) in cell lysates prepared using the Protein Expression Sample Preparation Kit. The assay probes consist of two oligonucleotides conjugated to antibodies that bind to separate epitopes of the hICAM1 protein. Key features of the TaqMan™ Protein Expression Assay:Specifications
Contains Assay Probe A, Assay Probe B, and Assay Probe Dilution Buffer. The assay probes (Assay Probe A and Assay Probe B) are supplied at a 20x concentration. The Assay Probe Dilution Buffer is supplied at a 1x concentration. Store at -15 to -25°C. | |
Low | |
AmpliTaq Fast DNA Polymerase | |
Proximity Ligation Assay | |
Sustainable packaging | |
ROX (Pre-mixed) | |
Protein Expression Assay Kit | |
Wet Ice | |
Target-Specific | |
100 Reactions |
Primer-probe | |
qPCR | |
Fast | |
7500 Fast System, 7900HT Fast System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode | |
FAM | |
TaqMan™ | |
UP (Ultra Pure) | |
100 Reactions | |
No | |
Cells |
For Research Use Only. Not for use in diagnostic procedures.