Learn More
Technical pH Buffer Solutions, METTLER TOLEDO™
Provides a reliable all-around pH buffer. Mettler Toledo™ Technical pH Buffer Solution are specified to two decimal places and are traceable to NIST standards. Color-coded solution comes in a handy 250mL plastic bottle with no-fuss labels.
Brand: Mettler Toledo 51350008
24.04 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : 59
Additional Details : Weight : 0.25920kg
Description
pH measurements are only as accurate as the buffer solutions used for calibration purposes. These technical-grade buffer solutions provide a reliable pH buffer for regular use with most applications. (pH electrodes should be calibrated at least once a day prior to measurements and after every cleaning, regeneration or long-term storage.)
- NIST traceable and specified to two decimal places
- Handy screw-top plastic bottles for easy handling
- Includes a test certificate and no-fuss label with a temperature table
- SDS and label contains information according to GHS (Globally Harmonized System)
Specifications
Blue | |
pH Electrode | |
250 mL | |
Plastic, screw-cap bottle | |
Technical |
pH measurement, electrode calibration | |
9.21 | |
Odorless | |
Yes |