Learn More
Medline™ Technico™ DIGIT-025 ATC Wine Pocket Refractometer
Brand: Medline™ 8145.0250M
Code : 03
Additional Details : Weight : 2.00000kg
Specifications
Wine Pocket Refractometer | |
DIGIT-025 ATC | |
1 Each |
0 to 25 Brix | |
Zero calibration with distilled water between 10 and 30°C. |