missing translation for 'onlineSavingsMsg'
Learn More

Tellurium(IV) bromide, 99.9% (metals basis)

Product Code. 11384879
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11384879

Brand: Thermo Scientific Alfa Aesar 075101.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10031-27-3 | Br4Te | 447.22 g/mol

Used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-27-3
Molecular Formula Br4Te
Molecular Weight (g/mol) 447.22
MDL Number MFCD00049575
InChI Key PTYIPBNVDTYPIO-UHFFFAOYSA-N
Synonym tellurium tetrabromide, tellurium iv bromide, tellurium bromide tebr4 , t-4, unii-7a29efj1af, 7a29efj1af, tetrabromo-, e4-tellane, tebr4, acmc-1bozh, tellurium iv bromide, ultra dry
PubChem CID 82311
IUPAC Name tetrabromo-λ⁴-tellane
SMILES Br[Te](Br)(Br)Br

Specifications

Boiling Point 420°C (decomposition)
Melting Point 380°C
Physical Form Crystalline
Quantity 50 g
UN Number UN2923
Merck Index 14,9125
Solubility Information Decomposes in water forming TeO2 and HBr
Formula Weight 447.27
Percent Purity 99.9%
Sensitivity Moisture sensitive
Density 4.31 g/mL at 15°C
Assay (metals basis)
Chemical Name or Material Tellurium (IV) bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tellurium bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Tellurium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.