missing translation for 'onlineSavingsMsg'
Learn More

tert-Butanol, ACS Reagent, ≥99.0%, Honeywell Riedel-de Haën™

Catalog Number p-7120858
Click to view available options
Quantity:
500 mL
1 L
2 L
20 L
Packaging:
Glass Bottle
Poly Bottle
Stainless Steel Can
This item is not returnable. View return policy
75-65-0
C4H10O
74.123
MFCD00004464
DKGAVHZHDRPRBM-UHFFFAOYSA-N
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
6386
CHEBI:45895
2-methylpropan-2-ol
CC(C)(C)O
This item is not returnable. View return policy
75-65-0
C4H10O
74.123
MFCD00004464
DKGAVHZHDRPRBM-UHFFFAOYSA-N
tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcohol
6386
CHEBI:45895
2-methylpropan-2-ol
CC(C)(C)O

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY
Melting Point 23°C to 26°C
Linear Formula (CH3)3COH
UN Number UN1120
Beilstein 906698
Formula Weight 74.12g/mol
Percent Purity ≥99%
Density 0.775g/mL at 25°C
Boiling Point 83°C
Quantity 500 mL
Explosion Limit 0.08
Flash Point 11°C (51.8°F)
Ignition Point 896°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.387
Vapor Pressure 44mmHg (26°C), 31mmHg (20°C)
Chemical Name or Material tert-Butanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'