missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate, ≥90%, Maybridge™

Brand:  Maybridge CC30013DA

Code : 31

Additional Details : CAS Number : 852180-47-3 Weight : 0.05100kg Transport : UN number : 3259 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 10027512

  • 138.56 € / 1g

Estimated Shipment
20-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

852180-47-3
291.395
TTXMFUXVXBAVIP-UHFFFAOYSA-N
2763841
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)CN
C16H25N3O2
MFCD03791213
1-boc-4-4-aminomethylphenyl piperazine, tert-butyl 4-4-aminomethyl phenyl piperazine-1-carboxylate, tert-butyl 4-4-aminomethyl phenyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-4-aminomethyl-phenyl-piperazine, 1-4-aminomethyl-phenyl-piperazine-4-carboxylic acid tert-butyl ester, 4-4-tert-butoxycarbonyl piperazin-1-yl benzylamine, n1-boc-4-aminomethyl phenylpiperazine, 4-aminomethyl phenylpiperazine, n1-boc protected, 1-4-aminomethyl-phenyl-piperazine-4-carboxylic a
tert-butyl 4-[4-(aminomethyl)phenyl]piperazine-1-carboxylate

Specifications

Tert-butyl 4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate
1g
89.5
90%
MFCD03791213
TTXMFUXVXBAVIP-UHFFFAOYSA-N
tert-butyl 4-[4-(aminomethyl)phenyl]piperazine-1-carboxylate
2763841
90%
Amber glass bottle
852180-47-3
100.0
C16H25N3O2
1-boc-4-4-aminomethylphenyl piperazine, tert-butyl 4-4-aminomethyl phenyl piperazine-1-carboxylate, tert-butyl 4-4-aminomethyl phenyl tetrahydro-1 2h-pyrazinecarboxylate, 1-boc-4-4-aminomethyl-phenyl-piperazine, 1-4-aminomethyl-phenyl-piperazine-4-carboxylic acid tert-butyl ester, 4-4-tert-butoxycarbonyl piperazin-1-yl benzylamine, n1-boc-4-aminomethyl phenylpiperazine, 4-aminomethyl phenylpiperazine, n1-boc protected, 1-4-aminomethyl-phenyl-piperazine-4-carboxylic a
CC(C)(C)OC(=O)N1CCN(CC1)C2=CC=C(C=C2)CN
291.395
291.39
Videos
SDS Documents